Assay Details
Target Gene Details
Entrez Gene ID: | 54108 |
Gene Name: | chromatin accessibility complex 1 |
Gene Aliases: |
CHARC1, CHARC15, CHRAC-1, CHRAC-15, CHRAC15, YCL1 |
Location: |
Chr.8:140511298-140517153 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRAC1 | NM_017444.5 | 3 | 1404 | NP_059140.1 |
| NR_023360.2 | 2 | 1277 | ||
| NR_040712.1 | 3 | 1099 | ||
| AK023537.1 | 3 | 1397 | BAB14601.1 | |
| AK308709.1 | 2 | 1147 | ||
| BC015891.1 | 3 | 1241 | AAH15891.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2737756 | Chr.8:140419230 - 141917755 on Build GRCh38 | Deletion |
|
| nsv509283 | Chr.8:140435015 - 140522401 on Build GRCh38 | Insertion |
|
| nsv1016282 | Chr.8:140453461 - 140537127 on Build GRCh38 | Gain |
|
| nsv1126125 | Chr.8:140419236 - 141917668 on Build GRCh38 | Deletion |
|
| dgv12367n54 | Chr.8:140431882 - 140524741 on Build GRCh38 | Gain |
|
| nsv465951 | Chr.8:140448395 - 140519958 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map