Assay Details
Target Gene Details
Entrez Gene ID: | 401541 |
Gene Name: | centromere protein P |
Gene Aliases: |
CENP-P |
Location: |
Chr.9:92325457-92615164 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPP | NM_001012267.2 | 8 | 2119 | NP_001012267.1 |
| NM_001286969.1 | 7 | 1937 | NP_001273898.1 | |
| NM_001286971.1 | 4 | 1276 | NP_001273900.1 | |
| AK091247.1 | 4 | 1277 | ||
| AK092603.1 | 8 | 1934 | ||
| AL833701.1 | 3 | 1794 | ||
| BX537851.1 | 7 | 1466 | CAD97859.1 |
Target Gene Details
Entrez Gene ID: | 64768 |
Gene Name: | inositol-pentakisphosphate 2-kinase |
Gene Aliases: |
C9orf12, INSP5K2, IP5K, IPK1, bA476B13.1 |
Location: |
Chr.9:92613184-92670265 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPPK | NM_022755.5 | 13 | 3715 | NP_073592.1 |
| AF351201.1 | 1 | 1295 | ||
| AK024267.1 | 11 | 3293 | BAB14866.1 |
More Information
Set Membership: |
|

Genomic Map