Assay Details
Target Gene Details
Entrez Gene ID: | 6886 |
Gene Name: | TAL bHLH transcription factor 1, erythroid differentiation factor |
Gene Aliases: |
SCL, TCL5, bHLHa17, tal-1 |
Location: |
Chr.1:47216290-47232389 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAL1 | NM_001287347.2 | NP_001274276.1 | ||
| NM_001290403.1 | NP_001277332.1 | |||
| NM_001290404.1 | NP_001277333.1 | |||
| NM_001290405.1 | NP_001277334.1 | |||
| NM_001290406.1 | NP_001277335.1 | |||
| NM_003189.5 | NP_003180.1 | |||
| XM_005271160.4 | XP_005271217.1 | |||
| XM_017002187.1 | XP_016857676.1 | |||
| XM_017002188.1 | XP_016857677.1 | |||
| XM_017002189.1 | XP_016857678.1 | |||
| XM_017002190.1 | XP_016857679.1 | |||
| XM_017002191.1 | XP_016857680.1 | |||
| XM_017002193.1 | XP_016857682.1 | |||
| M61108.1 | AAA36600.1 | |||
| S53245.1 | AAB19683.1 | |||
| S53686.1 | ||||
| X51990.1 | CAA36246.1 |
More Information
Set Membership: |
|

Genomic Map