Assay Details
Target Gene Details
Entrez Gene ID: | 1938 |
Gene Name: | eukaryotic translation elongation factor 2 |
Gene Aliases: |
EEF-2, EF-2, EF2, SCA26 |
Location: |
Chr.19:3976056-3985463 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EEF2 | NM_001961.3 | 12 | 1860 | NP_001952.1 |
| AK297398.1 | ||||
| AK298498.1 | 7 | 984 | ||
| AK299225.1 | ||||
| AY942181.1 | 11 | 1777 | AAX34409.1 | |
| BC024689.1 | 5 | 755 | AAH24689.1 | |
| BC126259.1 | 12 | 1829 | ||
| BC136313.1 | 12 | 1829 | ||
| M19997.1 | 3 | 279 | AAA50388.1 | |
| X51466.1 | 11 | 1777 | CAA35829.1 | |
| Z11692.1 | 11 | 1777 | CAA77750.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953949 | Chr.19:3959703 - 4089602 on Build GRCh38 | Deletion |
|
| esv1001715 | Chr.19:3972105 - 3981537 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map