Assay Details
Target Gene Details
Entrez Gene ID: | 9655 |
Gene Name: | suppressor of cytokine signaling 5 |
Gene Aliases: |
CIS6, CISH6, Cish5, SOCS-5 |
Location: |
Chr.2:46698960-46762788 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS5 | NM_014011.4 | 1 | 80 | NP_054730.1 |
| AB014571.1 | 1 | 88 | ||
| AK290194.1 | 1 | 94 | ||
| AK296796.1 | 1 | 91 | ||
| AL136896.1 | 1 | 80 | CAB66830.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1126548 | Chr.2:46698961 - 46699461 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map