Assay Details
Target Gene Details
Entrez Gene ID: | 5480 |
Gene Name: | peptidylprolyl isomerase C |
Gene Aliases: |
CYPC |
Location: |
Chr.5:123023383-123036730 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIC | NM_000943.4 | NP_000934.1 | ||
| AK290321.1 | ||||
| BC002678.2 | AAH02678.1 | |||
| BG721394.1 | ||||
| BU075157.1 | ||||
| BX472017.1 | ||||
| S71018.1 | AAB31350.1 |
Target Gene Details
Entrez Gene ID: | 28966 |
Gene Name: | sorting nexin 24 |
Gene Aliases: |
PRO1284, SBBI31 |
Location: |
Chr.5:122845465-123029318 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNX24 | XM_017009395.1 | XP_016864884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520349 | Chr.5:123023945 - 123120326 on Build GRCh38 | Loss |
|
| nsv1017500 | Chr.5:122899012 - 123100052 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map