Assay Details
Target Gene Details
Entrez Gene ID: | 9950 |
Gene Name: | golgin A5 |
Gene Aliases: |
GOLIM5, RFG5, ret-II |
Location: |
Chr.14:92794231-92839963 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGA5 | NM_005113.3 | 1 | 82 | NP_005104.3 |
| XM_011537420.2 | 1 | 82 | XP_011535722.1 | |
| AF085199.1 | AAD09753.1 | |||
| AK293077.1 | ||||
| AK303789.1 | ||||
| AL702150.1 | ||||
| BC023021.1 | AAH23021.1 | |||
| DB096772.1 | 1 | 82 |
More Information
Set Membership: |
|

Genomic Map