Assay Details
Target Gene Details
Entrez Gene ID: | 1544 |
Gene Name: | cytochrome P450 family 1 subfamily A member 2 |
Gene Aliases: |
CP12, P3-450, P450(PA) |
Location: |
Chr.15:74748843-74756600 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP1A2 | NM_000761.4 | 7 | 1472 | NP_000752.2 |
| AF182274.1 | 6 | 1416 | AAF13599.1 | |
| BC067424.1 | 7 | 1452 | AAH67424.1 | |
| BC067425.1 | 7 | 1452 | AAH67425.1 | |
| BC067426.1 | 7 | 1452 | AAH67426.1 | |
| BC067427.1 | 7 | 1452 | AAH67427.1 | |
| BC067428.1 | 7 | 1452 | AAH67428.1 | |
| BC067429.1 | 7 | 1452 | AAH67429.1 | |
| M12078.1 | 5 | 526 | AAA52154.1 | |
| M55053.1 | 6 | 1416 | AAA52146.1 | |
| Z00036.1 | 6 | 1416 | CAA77335.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833055 | Chr.15:74696003 - 74873947 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map