Assay Details
Target Gene Details
Entrez Gene ID: | 642 |
Gene Name: | bleomycin hydrolase |
Gene Aliases: |
BH, BMH |
Location: |
Chr.17:30248195-30292166 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLMH | NM_000386.3 | NP_000377.1 | ||
| AI283058.1 | ||||
| AK057600.1 | ||||
| BC003616.2 | AAH03616.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2023 | Chr.17:30245788 - 30260021 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map