Assay Details
Target Gene Details
Entrez Gene ID: | 4283 |
Gene Name: | C-X-C motif chemokine ligand 9 |
Gene Aliases: |
CMK, Humig, MIG, SCYB9, crg-10 |
Location: |
Chr.4:76001342-76007523 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL9 | NM_002416.2 | 1 | 104 | NP_002407.1 |
| AK311852.1 | 1 | 90 | ||
| BC063122.1 | 1 | 95 | AAH63122.1 | |
| BC095396.1 | 1 | 90 | AAH95396.1 | |
| DB141449.1 | 1 | 105 | ||
| X72755.1 | CAA51284.1 |
More Information
Set Membership: |
|

Genomic Map