Assay Details
Target Gene Details
Entrez Gene ID: | 8483 |
Gene Name: | cartilage intermediate layer protein |
Gene Aliases: |
CILP-1, HsT18872 |
Location: |
Chr.15:65195999-65211608 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CILP | NM_003613.3 | 9 | 2102 | NP_003604.3 |
| XM_017022678.1 | 8 | 2041 | XP_016878167.1 | |
| XM_017022679.1 | 8 | 2218 | XP_016878168.1 | |
| AF035408.1 | 9 | 2050 | AAC33838.1 | |
| AK225652.1 | ||||
| AK225926.1 | ||||
| AK313352.1 | 9 | 2073 | ||
| AL602624.1 | ||||
| AY358904.1 | 9 | 2056 | AAQ89263.1 | |
| BC035776.1 | 9 | 2049 | AAH35776.1 |
More Information
Set Membership: |
|

Genomic Map