Assay Details
Target Gene Details
Entrez Gene ID: | 84881 |
Gene Name: | RNA pseudouridylate synthase domain containing 4 |
Gene Aliases: |
- |
Location: |
Chr.11:126202094-126211692 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPUSD4 | NM_001144827.1 | 7 | 1551 | NP_001138299.1 |
| NM_032795.2 | 7 | 1644 | NP_116184.2 | |
| XM_011543039.2 | 7 | 1641 | XP_011541341.1 | |
| AK027400.1 | 7 | 1598 | BAB55086.1 | |
| AK300723.1 | 5 | 1118 | ||
| AK308059.1 | 7 | 1504 | ||
| BC014131.2 | 7 | 1605 | AAH14131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv536 | Chr.11:126163548 - 126207710 on Build GRCh38 | Deletion |
|
| nsv556528 | Chr.11:126185145 - 126210457 on Build GRCh38 | Loss |
|
| nsv556527 | Chr.11:126172311 - 126211508 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map