Assay Details
Target Gene Details
Entrez Gene ID: | 81894 |
Gene Name: | solute carrier family 25 member 28 |
Gene Aliases: |
MFRN2, MRS3/4, MRS4L, NPD016 |
Location: |
Chr.10:99610518-99659329 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A28 | NM_031212.3 | 4 | 1004 | NP_112489.3 |
| XM_005270208.1 | 4 | 994 | XP_005270265.1 | |
| XM_005270211.4 | 4 | 608 | XP_005270268.1 | |
| XM_006718005.3 | 4 | 668 | XP_006718068.1 | |
| XM_011540239.2 | 4 | 890 | XP_011538541.1 | |
| XM_011540240.1 | 5 | 2140 | XP_011538542.1 | |
| XM_017016741.1 | 4 | 2076 | XP_016872230.1 | |
| XM_017016742.1 | 4 | 1897 | XP_016872231.1 | |
| AF267854.1 | 1 | 691 | AAG44723.1 | |
| AF327402.1 | 4 | 904 | AAK49519.1 | |
| AF327403.1 | 4 | 1345 | AAK49520.1 | |
| AJ303077.1 | 4 | 904 | CAC27996.1 | |
| AJ303078.1 | 4 | 1345 | CAC27997.1 | |
| AK056782.1 | 1 | 1043 | ||
| AL831943.1 | 1 | 567 | CAH10774.1 | |
| BC047312.1 | 4 | 951 | AAH47312.1 | |
| BC058937.1 | 4 | 913 | AAH58937.1 | |
| BC076399.1 | 4 | 944 | AAH76399.1 | |
| BC094821.1 | 4 | 687 | AAH94821.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7515 | Chr.10:99606658 - 99639439 on Build GRCh38 | Insertion |
|
| nsv831958 | Chr.10:99451304 - 99651424 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map