Assay Details
Target Gene Details
Entrez Gene ID: | 151613 |
Gene Name: | tetratricopeptide repeat domain 14 |
Gene Aliases: |
DRDL5813, PRO19630 |
Location: |
Chr.3:180602130-180618347 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC14 | NM_001042601.2 | 1 | 172 | NP_001036066.1 |
| NM_001288582.1 | 1 | 172 | NP_001275511.1 | |
| NM_133462.3 | 1 | 172 | NP_597719.1 | |
| XM_017005739.1 | 1 | 159 | XP_016861228.1 | |
| XM_017005740.1 | 1 | 159 | XP_016861229.1 | |
| AK055984.1 | 1 | 130 | BAB71064.1 | |
| AL832052.1 | 1 | 110 | CAH10404.1 | |
| AL833086.2 | 1 | 109 | CAD89930.1 | |
| AL833103.1 | 1 | 91 | ||
| AY358762.1 | 1 | 92 | AAQ89122.1 | |
| BC027990.1 | 1 | 105 | AAH27990.1 | |
| BX474122.1 | 1 | 173 | ||
| BX648669.1 | 1 | 107 | ||
| DA283647.1 | 1 | 159 |
More Information
Set Membership: |
|

Genomic Map