Assay Details
Target Gene Details
Entrez Gene ID: | 8516 |
Gene Name: | integrin subunit alpha 8 |
Gene Aliases: |
- |
Location: |
Chr.10:15513949-15720335 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGA8 | NM_001291494.1 | NP_001278423.1 | ||
| NM_003638.2 | NP_003629.2 | |||
| AK297740.1 | ||||
| L36531.1 | AAA93514.1 |
More Information
Set Membership: |
|

Genomic Map