Assay Details
Target Gene Details
Entrez Gene ID: | 92579 |
Gene Name: | glucose-6-phosphatase catalytic subunit 3 |
Gene Aliases: |
SCN4, UGRP |
Location: |
Chr.17:44070700-44076344 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| G6PC3 | NM_001319945.1 | 1 | 178 | NP_001306874.1 |
| NM_138387.3 | 1 | 178 | NP_612396.1 | |
| NR_028581.1 | 1 | 178 | ||
| NR_028582.1 | 1 | 178 | ||
| XM_011525474.2 | 1 | 158 | XP_011523776.1 | |
| BC002494.2 | AAH02494.2 | |||
| BC021574.1 | 1 | 158 | AAH21574.1 | |
| BM790097.1 | 1 | 173 | ||
| DB460416.1 | 1 | 178 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833454 | Chr.17:43940848 - 44158311 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map