Assay Details
Target Gene Details
Entrez Gene ID: | 252969 |
Gene Name: | nei like DNA glycosylase 2 |
Gene Aliases: |
NEH2, NEI2 |
Location: |
Chr.8:11769663-11787346 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEIL2 | NM_001135746.1 | NP_001129218.1 | ||
| NM_001135747.1 | NP_001129219.1 | |||
| NM_001135748.1 | NP_001129220.1 | |||
| NM_145043.2 | NP_659480.1 | |||
| XM_005272382.2 | XP_005272439.1 | |||
| XM_005272383.2 | XP_005272440.1 | |||
| XM_017013301.1 | XP_016868790.1 | |||
| AK056206.1 | BAB71120.1 | |||
| AK097389.1 | BAC05030.1 | |||
| AK225837.1 | ||||
| AK294224.1 | ||||
| BC013952.2 | AAH13952.1 | |||
| BC013964.2 | AAH13964.1 | |||
| BX537529.1 | CAD97774.1 | |||
| DA205315.1 | ||||
| DA207321.1 | ||||
| DB452586.1 |
More Information
Set Membership: |
|

Genomic Map