Assay Details
Target Gene Details
Entrez Gene ID: | 83463 |
Gene Name: | MAX dimerization protein 3 |
Gene Aliases: |
BHLHC13, MAD3, MYX |
Location: |
Chr.5:177305500-177312291 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MXD3 | NM_001142935.1 | NP_001136407.1 | ||
| NM_031300.3 | NP_112590.1 | |||
| AF114834.1 | AAP97233.1 | |||
| AK057034.1 | BAB71352.1 | |||
| AK222579.1 | BAD96299.1 | |||
| AK303397.1 | ||||
| AK316468.1 | ||||
| AL522759.3 | ||||
| AL833959.1 | CAD38809.2 | |||
| BC000745.2 | AAH00745.1 | |||
| BC032586.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471057 | Chr.5:177310308 - 177469411 on Build GRCh38 | Loss |
|
| nsv509103 | Chr.5:177308751 - 177414575 on Build GRCh38 | Insertion |
|
| nsv600364 | Chr.5:177299634 - 177432847 on Build GRCh38 | Loss |
|
| nsv428132 | Chr.5:177191316 - 177375443 on Build GRCh38 | Gain |
|
| nsv1161311 | Chr.5:177305104 - 177546474 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map