Assay Details
Target Gene Details
Entrez Gene ID: | 9356 |
Gene Name: | solute carrier family 22 member 6 |
Gene Aliases: |
HOAT1, OAT1, PAHT, ROAT1 |
Location: |
Chr.11:62976597-62985023 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC22A6 | NM_004790.4 | NP_004781.2 | ||
| NM_153276.2 | NP_695008.1 | |||
| NM_153277.2 | NP_695009.1 | |||
| NM_153278.2 | NP_695010.1 | |||
| XM_017018562.1 | XP_016874051.1 | |||
| AB009697.1 | BAA75072.1 | |||
| AB009698.1 | BAA75073.1 | |||
| AF057039.2 | AAC70004.1 | |||
| AF097490.1 | AAD19356.1 | |||
| AF104038.1 | AAD10052.1 | |||
| AF124373.1 | AAD55356.1 | |||
| AJ251529.1 | CAB94830.1 | |||
| AJ271205.1 | CAB97249.1 | |||
| AK055764.1 | ||||
| AK091879.1 | ||||
| AK298374.1 | ||||
| BC033682.1 | AAH33682.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34011 | Chr.11:62885307 - 63226230 on Build GRCh38 | Loss |
|
| dgv204e214 | Chr.11:62950243 - 63024049 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map