Assay Details
Target Gene Details
Entrez Gene ID: | 670 |
Gene Name: | biphenyl hydrolase-like (serine hydrolase) |
Gene Aliases: |
BPH-RP, MCNAA, VACVASE |
Location: |
Chr.6:3118376-3153198 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPHL | NM_001302777.1 | NP_001289706.1 | ||
| NM_004332.3 | NP_004323.2 | |||
| NR_026648.2 | ||||
| NR_026649.2 | ||||
| AJ617684.1 | CAE85120.1 | |||
| AK297518.1 | ||||
| AK314909.1 | ||||
| AL832502.1 | ||||
| BC037778.1 | AAH37778.1 | |||
| BC106901.1 | AAI06902.1 | |||
| DC352501.1 | ||||
| X81372.1 | CAA57137.1 |
More Information
Set Membership: |
|

Genomic Map