Assay Details
Target Gene Details
Entrez Gene ID: | 4792 |
Gene Name: | NFKB inhibitor alpha |
Gene Aliases: |
IKBA, MAD-3, NFKBI |
Location: |
Chr.14:35401510-35404754 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFKBIA | NM_020529.2 | NP_065390.1 | ||
| AK311515.1 | ||||
| AK313421.1 | ||||
| AY033600.1 | AAK51149.1 | |||
| BC002601.2 | AAH02601.1 | |||
| BC004983.1 | AAH04983.1 | |||
| BT007091.1 | AAP35754.1 | |||
| M69043.1 | AAA16489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564320 | Chr.14:35402754 - 35406201 on Build GRCh38 | Loss |
|
| nsv564319 | Chr.14:35402011 - 35403586 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map