Assay Details
Target Gene Details
Entrez Gene ID: | 9862 |
Gene Name: | mediator complex subunit 24 |
Gene Aliases: |
ARC100, CRSP100, CRSP4, DRIP100, MED5, THRAP4, TRAP100 |
Location: |
Chr.17:40019097-40054636 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
Target Gene Details
Entrez Gene ID: | 102466757 |
Gene Name: | microRNA 6884 |
Gene Aliases: |
hsa-mir-6884 |
Location: |
Chr.17:40026332-40026409 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6884 | NR_106944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573064 | Chr.17:39896348 - 40029806 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map