Assay Details
Target Gene Details
Entrez Gene ID: | 6612 |
Gene Name: | small ubiquitin-like modifier 3 |
Gene Aliases: |
SMT3A, SMT3H1, SUMO-3, Smt3B |
Location: |
Chr.21:44805617-44818129 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUMO3 | NM_001286416.1 | 4 | 540 | NP_001273345.1 |
| NM_006936.2 | 4 | 426 | NP_008867.2 | |
| AK222591.1 | 4 | 357 | BAD96311.1 | |
| AK300822.1 | 4 | 492 | ||
| AK312350.1 | ||||
| BC000036.2 | 4 | 351 | AAH00036.1 | |
| BC008420.1 | 4 | 426 | AAH08420.1 | |
| BT007008.1 | AAP35654.1 | |||
| CR542173.1 | CAG46970.1 | |||
| CR542188.1 | CAG46985.1 | |||
| X99584.1 | 4 | 359 | CAA67896.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647136 | Chr.21:44802580 - 44905193 on Build GRCh38 | Loss |
|
| esv3647135 | Chr.21:44764118 - 44826199 on Build GRCh38 | Loss |
|
| nsv459298 | Chr.21:44771532 - 44858053 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map