Assay Details
Target Gene Details
Entrez Gene ID: | 9550 |
Gene Name: | ATPase H+ transporting V1 subunit G1 |
Gene Aliases: |
ATP6G, ATP6G1, ATP6GL, ATP6J, Vma10 |
Location: |
Chr.9:114587714-114598872 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V1G1 | NM_004888.3 | 3 | 1230 | NP_004879.1 |
| AI092246.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv12859n54 | Chr.9:114193988 - 114696195 on Build GRCh38 | Gain |
|
| dgv7706n100 | Chr.9:114182745 - 114697870 on Build GRCh38 | Gain |
|
| nsv831694 | Chr.9:114566360 - 114740505 on Build GRCh38 | Gain |
|
| dgv33e196 | Chr.9:114030664 - 114604363 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116299071] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map