Assay Details
Target Gene Details
Entrez Gene ID: | 6605 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1 |
Gene Aliases: |
BAF57, CSS5 |
Location: |
Chr.17:40627720-40647851 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMARCE1 | NM_003079.4 | 11 | 1525 | NP_003070.3 |
| AK095047.1 | 11 | 1492 | ||
| AK294218.1 | 8 | 1311 | ||
| AK294666.1 | 9 | 1417 | ||
| BC063700.1 | 11 | 1488 | AAH63700.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574994 | Chr.17:40563547 - 40668384 on Build GRCh38 | Gain |
|
| esv2758689 | Chr.17:40592048 - 40693852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map