Assay Details
Target Gene Details
Entrez Gene ID: | 53343 |
Gene Name: | nudix hydrolase 9 |
Gene Aliases: |
NUDT10 |
Location: |
Chr.4:87420002-87459454 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT9 | NM_001248011.1 | NP_001234940.1 | ||
| NM_024047.4 | NP_076952.1 | |||
| NM_198038.2 | NP_932155.1 | |||
| XM_011532031.2 | XP_011530333.1 | |||
| XM_011532032.2 | XP_011530334.1 | |||
| AF265341.1 | AAM46066.1 | |||
| AF273028.1 | AAM46068.1 | |||
| AK027295.1 | BAB55021.1 | |||
| AK074845.1 | BAC11239.1 | |||
| AY026252.1 | AAK07671.1 | |||
| BM544951.1 | ||||
| BP277630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004518 | Chr.4:87297321 - 87433251 on Build GRCh38 | Gain |
|
| esv3601274 | Chr.4:87364198 - 87422760 on Build GRCh38 | Loss |
|
| nsv1002789 | Chr.4:87318188 - 87685333 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map