Assay Details
Target Gene Details
Entrez Gene ID: | 200014 |
Gene Name: | coiled-coil and C2 domain containing 1B |
Gene Aliases: |
- |
Location: |
Chr.1:52348437-52366291 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CC2D1B | NM_032449.2 | 24 | 5044 | NP_115825.1 |
| XM_005270590.2 | 25 | 4836 | XP_005270647.1 | |
| AB058739.1 | 20 | 4475 | BAB47465.1 | |
| AK095487.1 | 1 | 1578 |
More Information
Set Membership: |
|

Genomic Map