Assay Details
Target Gene Details
Entrez Gene ID: | 83884 |
Gene Name: | solute carrier family 25 member 2 |
Gene Aliases: |
ORC2, ORNT2 |
Location: |
Chr.5:141302629-141304063 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A2 | NM_031947.3 | 1 | 1169 | NP_114153.1 |
| AF332005.1 | 1 | 971 | AAK26320.1 | |
| AF378119.1 | 1 | 1151 | AAM94902.1 | |
| AY208943.1 | 1 | 1079 | AAO31753.1 | |
| BC069458.1 | AAH69458.1 | |||
| BC100935.2 | AAI00936.1 | |||
| BC100936.1 | AAI00937.1 | |||
| BC100937.2 | AAI00938.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759382 | Chr.5:140777355 - 141449115 on Build GRCh38 | Gain |
|
| nsv428127 | Chr.5:141016768 - 141319173 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map