Assay Details
Target Gene Details
Entrez Gene ID: | 2788 |
Gene Name: | G protein subunit gamma 7 |
Gene Aliases: |
- |
Location: |
Chr.19:2511219-2702748 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNG7 | NM_052847.2 | NP_443079.1 | ||
| XM_017026606.1 | XP_016882095.1 | |||
| AB010414.1 | BAA28348.1 | |||
| AF493874.1 | AAM12588.1 | |||
| AK024465.1 | BAB15755.1 | |||
| AK311750.1 | ||||
| BC014466.1 | AAH14466.1 | |||
| BC053630.1 | AAH53630.1 | |||
| DA106844.1 | ||||
| DA247608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064930 | Chr.19:2380093 - 2592252 on Build GRCh38 | Gain |
|
| esv2762019 | Chr.19:2503630 - 2519855 on Build GRCh38 | Gain |
|
| nsv953944 | Chr.19:2283702 - 2525802 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map