Assay Details
Target Gene Details
Entrez Gene ID: | 57701 |
Gene Name: | NCK associated protein 5 like |
Gene Aliases: |
Cep169, FP1193, KIAA1602 |
Location: |
Chr.12:49791139-49828750 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCKAP5L | NM_001037806.3 | NP_001032895.2 | ||
| XM_005269050.3 | XP_005269107.1 | |||
| XM_006719525.2 | XP_006719588.1 | |||
| XM_011538610.2 | XP_011536912.1 | |||
| XM_011538611.2 | XP_011536913.1 | |||
| AK093900.1 | 3 | 1979 | ||
| BC110599.2 | 2 | 1750 | AAI10600.1 | |
| CN359946.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522972 | Chr.12:49802611 - 50017048 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map