Assay Details
Target Gene Details
Entrez Gene ID: | 55702 |
Gene Name: | coiled-coil domain containing 94 |
Gene Aliases: |
- |
Location: |
Chr.19:4247114-4269088 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC94 | NM_018074.4 | NP_060544.2 | ||
| AF091080.1 | AAC72949.1 | |||
| AK001236.1 | BAA91572.1 | |||
| AK023988.1 | BAB14757.1 | |||
| BC000561.2 | AAH00561.1 | |||
| BC019096.2 | AAH19096.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578442 | Chr.19:4236999 - 4312106 on Build GRCh38 | Loss |
|
| nsv833717 | Chr.19:4204757 - 4368519 on Build GRCh38 | Loss |
|
| nsv833718 | Chr.19:4221577 - 4480248 on Build GRCh38 | Loss |
|
| nsv833716 | Chr.19:4121259 - 4274854 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map