Assay Details
Target Gene Details
Entrez Gene ID: | 140679 |
Gene Name: | solute carrier family 32 member 1 |
Gene Aliases: |
VGAT, VIAAT |
Location: |
Chr.20:38724462-38729372 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC32A1 | NM_080552.2 | 2 | 1761 | NP_542119.1 |
| AK055051.1 | 2 | 1737 | BAB70846.1 | |
| AK091079.1 | 2 | 1245 | ||
| AY044836.1 | 2 | 1503 | AAK98782.1 | |
| BC036458.2 | 2 | 1348 | AAH36458.2 | |
| BC053582.1 | 2 | 1672 | AAH53582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058457 | Chr.20:38277492 - 38844429 on Build GRCh38 | Loss |
|
| nsv954035 | Chr.20:38722258 - 38731557 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map