Assay Details
Target Gene Details
Entrez Gene ID: | 54872 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class G |
Gene Aliases: |
GPI7, LAS21, MRT53, PRO4405, RLGS1930 |
Location: |
Chr.4:499200-539921 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGG | NM_001127178.2 | 1 | 147 | NP_001120650.1 |
| NM_001289051.1 | NP_001275980.1 | |||
| NM_001289052.1 | 1 | 147 | NP_001275981.1 | |
| NM_001289053.1 | 1 | 147 | NP_001275982.1 | |
| NM_001289055.1 | 1 | 147 | NP_001275984.1 | |
| NM_001289057.1 | 1 | 147 | NP_001275986.1 | |
| NM_017733.4 | 1 | 147 | NP_060203.3 | |
| NR_110293.1 | 1 | 147 | ||
| XM_005272284.2 | 1 | 145 | XP_005272341.1 | |
| XM_011513490.2 | 1 | 110 | XP_011511792.1 | |
| XM_011513491.1 | 1 | 107 | XP_011511793.1 | |
| XM_011513493.1 | 1 | 147 | XP_011511795.1 | |
| XM_011513494.2 | 1 | 147 | XP_011511796.1 | |
| XM_017008321.1 | 1 | 145 | XP_016863810.1 | |
| XM_017008322.1 | 1 | 145 | XP_016863811.1 | |
| XM_017008323.1 | 1 | 145 | XP_016863812.1 | |
| XM_017008324.1 | 1 | 145 | XP_016863813.1 | |
| XM_017008325.1 | 1 | 94 | XP_016863814.1 | |
| XM_017008326.1 | 1 | 147 | XP_016863815.1 | |
| AB162713.1 | 1 | 117 | BAD89023.1 | |
| AK074715.1 | BAC11157.1 | |||
| AK074815.1 | 1 | 126 | BAC11227.1 | |
| AK097244.1 | 1 | 107 | BAC04984.1 | |
| AK296507.1 | 1 | 138 | ||
| AK309107.1 | 1 | 107 | ||
| AY358538.1 | 1 | 89 | AAQ88902.1 | |
| BC110878.1 | 1 | 89 | AAI10879.1 | |
| DA548833.1 | 1 | 147 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599372 | Chr.4:473707 - 505788 on Build GRCh38 | Loss |
|
| nsv1001899 | Chr.4:68917 - 523947 on Build GRCh38 | Loss |
|
| nsv593202 | Chr.4:421522 - 503349 on Build GRCh38 | Gain |
|
| esv3599371 | Chr.4:442079 - 503495 on Build GRCh38 | Loss |
|
| nsv1006781 | Chr.4:343385 - 560798 on Build GRCh38 | Gain |
|
| esv3893859 | Chr.4:416226 - 503349 on Build GRCh38 | Gain |
|
| nsv593204 | Chr.4:476314 - 534500 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map