Assay Details
Target Gene Details
Entrez Gene ID: | 8916 |
Gene Name: | HECT and RLD domain containing E3 ubiquitin protein ligase 3 |
Gene Aliases: |
- |
Location: |
Chr.4:88592423-88709303 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HERC3 | NM_001271602.1 | NP_001258531.1 | ||
| NM_014606.2 | NP_055421.1 | |||
| XM_005263327.3 | XP_005263384.1 | |||
| XM_005263329.3 | XP_005263386.1 | |||
| XM_005263330.3 | XP_005263387.1 | |||
| XM_011532397.2 | XP_011530699.1 | |||
| XM_017008806.1 | XP_016864295.1 | |||
| XM_017008807.1 | XP_016864296.1 | |||
| XM_017008808.1 | XP_016864297.1 | |||
| AK289990.1 | ||||
| AK296379.1 | ||||
| AK296397.1 | ||||
| D25215.1 |
Target Gene Details
Entrez Gene ID: | 266812 |
Gene Name: | nucleosome assembly protein 1 like 5 |
Gene Aliases: |
DRLM |
Location: |
Chr.4:88695913-88698235 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAP1L5 | NM_153757.3 | 1 | 1330 | NP_715638.1 |
| AK054689.1 | 1 | 924 | BAB70793.1 | |
| AL834329.1 | 1 | 909 | CAD38997.1 | |
| BC104883.1 | 1 | 873 | AAI04884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461576 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map