Assay Details
Target Gene Details
Entrez Gene ID: | 6275 |
Gene Name: | S100 calcium binding protein A4 |
Gene Aliases: |
18A2, 42A, CAPL, FSP1, MTS1, P9KA, PEL98 |
Location: |
Chr.1:153543619-153545806 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A4 | NM_002961.2 | 3 | 331 | NP_002952.1 |
| NM_019554.2 | 4 | 380 | NP_062427.1 | |
| AK292083.1 | 3 | 331 | ||
| AV735911.1 | 2 | 214 | ||
| BC000838.1 | 4 | 364 | AAH00838.1 | |
| BC016300.1 | 3 | 335 | AAH16300.1 | |
| CB109749.1 | 4 | 380 | ||
| CR450345.1 | CAG29341.1 | |||
| M80563.1 | 3 | 397 | AAA51920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831603 | Chr.1:153433366 - 153603667 on Build GRCh38 | Gain |
|
| nsv1002705 | Chr.1:153335376 - 153553113 on Build GRCh38 | Gain |
|
| esv3587591 | Chr.1:153528630 - 153570193 on Build GRCh38 | Gain |
|
| esv3587584 | Chr.1:153466126 - 153598991 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map