Assay Details
Target Gene Details
Entrez Gene ID: | 28990 |
Gene Name: | asteroid homolog 1 (Drosophila) |
Gene Aliases: |
HT001 |
Location: |
Chr.3:131013873-131026854 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASTE1 | NM_001288950.1 | 4 | 1660 | NP_001275879.1 |
NM_014065.3 | 4 | 1885 | NP_054784.2 | |
NR_110229.1 | 4 | 1885 | ||
XM_017006266.1 | 4 | 1801 | XP_016861755.1 | |
XM_017006267.1 | 4 | 1959 | XP_016861756.1 | |
XM_017006268.1 | 4 | 1880 | XP_016861757.1 | |
XM_017006269.1 | 4 | 1801 | XP_016861758.1 | |
AF113539.1 | 2 | 555 | AAF14876.1 | |
AK027258.1 | 2 | 1275 | ||
AK027806.1 | 4 | 1833 | BAB55383.1 | |
AK294156.1 | 4 | 1885 | ||
AK301388.1 | ||||
BC030274.1 | 2 | 505 | ||
BC103709.1 | 2 | 976 | AAI03710.1 | |
BC110616.2 | 4 | 1598 | AAI10617.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
