Assay Details
Target Gene Details
Entrez Gene ID: | 150737 |
Gene Name: | tetratricopeptide repeat domain 30B |
Gene Aliases: |
IFT70, IFT70B, fleer |
Location: |
Chr.2:177550152-177552796 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC30B | NM_152517.2 | 1 | 2288 | NP_689730.2 |
| AI498353.1 | ||||
| AK055552.1 | BAB70953.1 | |||
| AK296828.1 | 2 | 2125 | ||
| AW007843.1 | 1 | 368 | ||
| BX647620.1 | CAH56200.1 |
More Information
Set Membership: |
|

Genomic Map