Assay Details
Target Gene Details
Entrez Gene ID: | 51652 |
Gene Name: | charged multivesicular body protein 3 |
Gene Aliases: |
CGI-149, NEDF, VPS24 |
Location: |
Chr.2:86503430-86563497 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHMP3 | NM_001005753.2 | NP_001005753.1 | ||
| NM_001193517.1 | NP_001180446.1 | |||
| NM_016079.3 | NP_057163.1 | |||
| NR_036454.1 | ||||
| AF151907.1 | AAD34144.1 | |||
| AF219226.1 | AAF26737.1 | |||
| AK025260.1 | ||||
| AK095433.1 | ||||
| AK129502.1 | ||||
| AK290725.1 | ||||
| AK312353.1 | ||||
| AK315835.1 | ||||
| AY364249.1 | AAQ76808.1 | |||
| BC004419.1 | AAH04419.1 | |||
| BG705423.1 | ||||
| BI562428.1 | ||||
| DA046087.1 | ||||
| DA290886.1 |
Target Gene Details
Entrez Gene ID: | 100526767 |
Gene Name: | RNF103-CHMP3 readthrough |
Gene Aliases: |
RNF103-VPS24 |
Location: |
Chr.2:86503430-86721122 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF103-CHMP3 | NM_001198954.1 | NP_001185883.1 | ||
| AK294389.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2813 | Chr.2:86480671 - 86514180 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map