Assay Details
Target Gene Details
Entrez Gene ID: | 11198 |
Gene Name: | SPT16 homolog, facilitates chromatin remodeling subunit |
Gene Aliases: |
CDC68, FACTP140, SPT16, SPT16/CDC68 |
Location: |
Chr.14:21351472-21384266 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUPT16H | NM_007192.3 | NP_009123.1 | ||
| XM_011536381.2 | XP_011534683.1 | |||
| AK001719.1 | ||||
| AK024072.1 | ||||
| BC000565.2 | AAH00565.1 | |||
| BC014046.2 | AAH14046.1 | |||
| BC064561.1 | AAH64561.1 | |||
| BC073849.1 | AAH73849.1 |
More Information
Set Membership: |
|

Genomic Map