Assay Details
Target Gene Details
Entrez Gene ID: | 23016 |
Gene Name: | exosome component 7 |
Gene Aliases: |
EAP1, RRP42, Rrp42p, hRrp42p, p8 |
Location: |
Chr.3:44975291-45012668 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOSC7 | NM_015004.3 | NP_055819.2 | ||
| NR_023353.1 | ||||
| XM_017005930.1 | XP_016861419.1 | |||
| AK130053.1 | ||||
| AK295779.1 | ||||
| AK315741.1 | ||||
| BC012831.2 | AAH12831.1 | |||
| BU153856.1 | ||||
| D29958.1 | BAA06226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834676 | Chr.3:44928707 - 45149356 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map