Assay Details
Target Gene Details
Entrez Gene ID: | 2323 |
Gene Name: | fms related tyrosine kinase 3 ligand |
Gene Aliases: |
FL, FLT3L |
Location: |
Chr.19:49474172-49487038 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv833862 | Chr.19:49448789 - 49570676 on Build GRCh38 | Loss |
|
| esv3644624 | Chr.19:49461676 - 49475440 on Build GRCh38 | Gain |
|
| nsv953599 | Chr.19:49386444 - 49506743 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75750998] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map