Assay Details
Target Gene Details
Entrez Gene ID: | 27 |
Gene Name: | ABL proto-oncogene 2, non-receptor tyrosine kinase |
Gene Aliases: |
ABLL, ARG |
Location: |
Chr.1:179099327-179229693 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABL2 | NM_001136000.2 | 13 | 2238 | NP_001129472.1 |
NM_001168236.1 | 11 | 2771 | NP_001161708.1 | |
NM_001168237.1 | 13 | 2525 | NP_001161709.1 | |
NM_001168238.1 | 12 | 2462 | NP_001161710.1 | |
NM_001168239.1 | 12 | 2175 | NP_001161711.1 | |
NM_005158.4 | 12 | 2547 | NP_005149.4 | |
NM_007314.3 | 12 | 2834 | NP_009298.1 | |
XM_005245088.2 | 11 | 2469 | XP_005245145.1 | |
XM_017001035.1 | 11 | 5144 | XP_016856524.1 | |
BC065912.1 | 12 | 2514 | AAH65912.1 | |
BX538317.1 | 13 | 2277 | CAD98092.1 | |
FJ542283.1 | 12 | 2130 | ||
FJ542284.1 | 13 | 2238 | ||
FJ542285.1 | 11 | 2484 | ||
FJ542286.1 | 12 | 2175 | ||
M35296.1 | 12 | 2751 | AAA35553.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
