Assay Details
Target Gene Details
Entrez Gene ID: | 5832 |
Gene Name: | aldehyde dehydrogenase 18 family member A1 |
Gene Aliases: |
ADCL3, ARCL3A, GSAS, P5CS, PYCS, SPG9A, SPG9B |
Location: |
Chr.10:95605929-95656810 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH18A1 | NM_001017423.1 | 18 | 2748 | NP_001017423.1 |
| NM_001323412.1 | 17 | 2539 | NP_001310341.1 | |
| NM_001323413.1 | 18 | 2905 | NP_001310342.1 | |
| NM_001323414.1 | 18 | 2774 | NP_001310343.1 | |
| NM_001323415.1 | 18 | 2899 | NP_001310344.1 | |
| NM_001323416.1 | 17 | 2690 | NP_001310345.1 | |
| NM_001323417.1 | 17 | 2649 | NP_001310346.1 | |
| NM_001323418.1 | 17 | 2684 | NP_001310347.1 | |
| NM_001323419.1 | 15 | 2284 | NP_001310348.1 | |
| NM_002860.3 | 18 | 2754 | NP_002851.2 | |
| AK126355.1 | 1 | 3135 | ||
| AK225455.1 | ||||
| AK225465.1 | ||||
| AK226032.1 | ||||
| BC106054.1 | 18 | 2589 | AAI06055.1 | |
| BC117240.1 | 18 | 2695 | ||
| BC117242.1 | 18 | 2695 | ||
| BC143930.1 | 18 | 2689 | ||
| BU170321.1 | ||||
| U68758.1 | 18 | 2563 | AAD00169.1 | |
| U76542.1 | 18 | 2569 | AAD17454.1 | |
| X94453.1 | 18 | 2650 | CAA64224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526180 | Chr.10:95572438 - 95751280 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map