Assay Details
Target Gene Details
Entrez Gene ID: | 51067 |
Gene Name: | tyrosyl-tRNA synthetase 2 |
Gene Aliases: |
CGI-04, MLASA2, MT-TYRRS, TYRRS |
Location: |
Chr.12:32725247-32755961 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YARS2 | NM_001040436.2 | 5 | 1825 | NP_001035526.1 |
| AF132939.1 | 5 | 1748 | AAD27714.1 | |
| AI861961.1 | 1 | 349 | ||
| BM979973.1 | 1 | 435 | ||
| EB386795.1 | 1 | 316 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1429n100 | Chr.12:32718926 - 32775899 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map