Assay Details
Target Gene Details
Entrez Gene ID: | 10549 |
Gene Name: | peroxiredoxin 4 |
Gene Aliases: |
AOE37-2, AOE372, HEL-S-97n, PRX-4 |
Location: |
Chr.X:23664260-23686399 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRDX4 | NM_006406.1 | NP_006397.1 | ||
| XM_005274438.1 | XP_005274495.1 | |||
| XM_017029230.1 | XP_016884719.1 | |||
| XM_017029231.1 | XP_016884720.1 | |||
| BC003609.2 | AAH03609.1 | |||
| BC007107.1 | AAH07107.1 | |||
| BC016770.1 | AAH16770.1 | |||
| CR541668.1 | CAG46469.1 | |||
| CR541705.1 | CAG46506.1 | |||
| FJ224297.1 | ||||
| U25182.1 | AAB95175.1 |
More Information
Set Membership: |
|

Genomic Map