Assay Details
Target Gene Details
Entrez Gene ID: | 80347 |
Gene Name: | Coenzyme A synthase |
Gene Aliases: |
DPCK, NBIA6, NBP, PPAT, UKR1, pOV-2 |
Location: |
Chr.17:42562074-42566281 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COASY | NM_001042529.2 | 10 | 2012 | NP_001035994.1 |
| NM_001042532.3 | 11 | 2144 | NP_001035997.2 | |
| NM_025233.6 | 9 | 2404 | NP_079509.5 | |
| XM_006722116.3 | 10 | 2038 | XP_006722179.1 | |
| XM_011525300.1 | 10 | 2208 | XP_011523602.1 | |
| XM_017025167.1 | 7 | 2093 | XP_016880656.1 | |
| XM_017025168.1 | 10 | 1617 | XP_016880657.1 | |
| AF208536.1 | 9 | 1364 | AAF87955.1 | |
| AK075415.1 | 9 | 2312 | ||
| AK092498.1 | 7 | 2079 | ||
| AK225485.1 | 10 | 1941 | ||
| AL360167.1 | 9 | 1364 | ||
| AL360168.1 | 3 | 791 | ||
| AL833588.1 | 10 | 1936 | ||
| AY094602.1 | 10 | 1965 | AAM19996.1 | |
| BC006354.1 | 8 | 1105 | AAH06354.1 | |
| BC020985.1 | 9 | 1580 | AAH20985.1 | |
| BC067254.1 | 9 | 2308 | AAH67254.1 | |
| U18919.1 | 7 | 863 | AAA69699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064012 | Chr.17:42427786 - 42606703 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map