Assay Details
Target Gene Details
Entrez Gene ID: | 51147 |
Gene Name: | inhibitor of growth family member 4 |
Gene Aliases: |
my036, p29ING4 |
Location: |
Chr.12:6650538-6663143 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117962564] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map