Assay Details
Target Gene Details
Entrez Gene ID: | 56034 |
Gene Name: | platelet derived growth factor C |
Gene Aliases: |
FALLOTEIN, SCDGF |
Location: |
Chr.4:156761611-156971394 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDGFC | NM_016205.2 | 6 | 2150 | NP_057289.1 |
| NR_036641.1 | 7 | 2211 | ||
| XM_011532124.2 | 8 | 1830 | XP_011530426.1 | |
| XM_017008455.1 | 6 | 1779 | XP_016863944.1 | |
| XM_017008456.1 | 6 | 1923 | XP_016863945.1 | |
| AF091434.1 | 6 | 2150 | AAF00049.1 | |
| AF244813.1 | 6 | 1847 | AAF80597.1 | |
| AF260738.1 | AAK51637.1 | |||
| AY358493.1 | 6 | 1943 | AAQ88857.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830122 | Chr.4:156695746 - 156892693 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map