Assay Details
Target Gene Details
Entrez Gene ID: | 127733 |
Gene Name: | UBX domain protein 10 |
Gene Aliases: |
UBXD3 |
Location: |
Chr.1:20183106-20196060 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBXN10 | NM_152376.4 | 2 | 1392 | NP_689589.1 |
| XM_005245742.3 | 2 | 1896 | XP_005245799.1 | |
| XM_011540699.2 | 2 | 1600 | XP_011539001.1 | |
| AK058158.1 | BAB71693.1 | |||
| BC036417.1 | 1 | 802 | AAH36417.1 | |
| BC047428.1 | 2 | 1387 | AAH47428.1 | |
| BX648631.1 | 2 | 1382 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|

Genomic Map