Assay Details
Target Gene Details
Entrez Gene ID: | 6749 |
Gene Name: | structure specific recognition protein 1 |
Gene Aliases: |
FACT, FACT80, T160 |
Location: |
Chr.11:57325985-57335877 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSRP1 | NM_003146.2 | NP_003137.1 | ||
| XM_017018180.1 | XP_016873669.1 | |||
| AB209132.1 | BAD92369.1 | |||
| BC005116.1 | AAH05116.1 | |||
| M86737.1 | AAA58660.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74516355] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map